Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001020676.1 | NP_001018512.1 | 3 - 4 | 338 | 64 | |
GenBank mRNA | BC095734.1 | - | 3 - 4 | 338 | 64 | |
BC155231.1 | - | 3 - 4 | 338 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |