Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trpc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001030166.1 | NP_001025337.1 | 7 - 8 | 1632 | 81 | |
GenBank mRNA | AY974804.1 | - | 7 - 8 | 1632 | 81 | |
BC163641.1 | - | 7 - 8 | 1716 | 81 | ||
KF446628.1 | - | 7 - 8 | 1665 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |