Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sla1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034989.2 | NP_001030161.1 | 2 - 3 | 245 | 69 | |
GenBank mRNA | BC103485.1 | - | 2 - 3 | 249 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |