Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: etsrp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001037375.1 | NP_001032452.1 | 3 - 4 | 246 | 63 | |
XM_005158213.4 | XP_005158270.1 | 3 - 4 | 478 | 63 | ||
GenBank mRNA | BC124743.1 | - | 3 - 4 | 255 | 63 | |
BC164765.1 | - | 2 - 3 | 216 | 63 | ||
DQ021472.1 | - | 3 - 4 | 246 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |