Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ripply2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039109.1 | NP_001034198.1 | 1 - 2 | 156 | 114 | |
GenBank mRNA | AB212220.1 | - | 1 - 2 | 156 | 114 | |
BC162794.1 | - | 1 - 2 | 130 | 114 | ||
BC162809.1 | - | 1 - 2 | 130 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |