Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: illr4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040040.1 | NP_001035129.1 | 1 - 2 | 102 | 75 | |
GenBank mRNA | AY986758.1 | - | 1 - 2 | 102 | 75 | |
BC124080.1 | - | 1 - 2 | 128 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |