Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sult4a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040244.1 | NP_001035334.1 | 5 - 6 | 689 | 74 | |
GenBank mRNA | BC115282.1 | - | 5 - 6 | 689 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |