Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: map4k2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001040358.1 | NP_001035448.1 | 5 - 6 | 464 | 124 | |
GenBank mRNA | BC115221.1 | - | 5 - 6 | 464 | 124 | |
BC162619.1 | - | 5 - 6 | 562 | 124 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |