Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mov10b.1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044342.2 | NP_001037807.2 | 2 - 3 | 282 | 74 | |
GenBank mRNA | BC171375.1 | - | 2 - 3 | 282 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |