Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rnd1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044396.2 | NP_001037861.1 | 2 - 3 | 345 | 88 | |
GenBank mRNA | AY865571.1 | - | 2 - 3 | 203 | 88 | |
BC122138.1 | - | 2 - 3 | 345 | 88 | ||
BC165155.1 | - | 2 - 3 | 237 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |