Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btbd11a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045002.1 | NP_001038467.1 | 8 - 9 | 1030 | 71 | |
NM_001327842.1 | NP_001314771.1 | 9 - 10 | 2239 | 71 | ||
XM_005164650.4 | XP_005164707.1 | 8 - 9 | 1861 | 71 | ||
XM_009300161.2 | XP_009298436.1 | 8 - 9 | 1027 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |