Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045044.1 | NP_001038509.1 | 11 - 12 | 1098 | 93 | |
XM_005161685.4 | XP_005161742.1 | 11 - 12 | 1160 | 93 | ||
XM_009296204.3 | XP_009294479.1 | 11 - 12 | 1167 | 93 | ||
XM_021469341.1 | XP_021325016.1 | 5 - 6 | 3464 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Recommended Assay Danio rerio Probe spans exons |