Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glsa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045044.1 | NP_001038509.1 | 8 - 9 | 921 | 109 | |
XM_005161685.4 | XP_005161742.1 | 8 - 9 | 983 | 109 | ||
XM_009296204.3 | XP_009294479.1 | 8 - 9 | 990 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |