Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otomp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045087.1 | NP_001038552.1 | 6 - 7 | 750 | 82 | |
GenBank mRNA | AY681353.1 | - | 6 - 7 | 750 | 82 | |
BC076449.1 | - | 6 - 7 | 82 | |||
BC134025.1 | - | 6 - 7 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |