Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpreb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045177.1 | NP_001038642.1 | 7 - 8 | 731 | 82 | |
XM_005158789.3 | XP_005158846.1 | 7 - 8 | 774 | 82 | ||
XM_017351664.2 | XP_017207153.1 | 4 - 5 | 463 | 82 | ||
GenBank mRNA | BC128883.1 | - | 7 - 8 | 732 | 82 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |