Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cgrrf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045257.1 | NP_001038722.1 | 5 - 6 | 777 | 71 | |
GenBank mRNA | BC116465.1 | - | 5 - 6 | 777 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |