Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnk1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001080017.2 | NP_001073486.2 | 11 - 12 | 1542 | 71 | |
XM_005172298.4 | XP_005172355.1 | 11 - 12 | 1760 | 71 | ||
XM_017356886.2 | XP_017212375.1 | 10 - 11 | 1662 | 71 | ||
GenBank mRNA | BC125935.1 | - | 11 - 12 | 1542 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |