Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082839.1 | NP_001076308.1 | 1 - 2 | 153 | 88 | |
XM_005165174.4 | XP_005165231.1 | 1 - 2 | 440 | 88 | ||
XM_009301350.3 | XP_009299625.1 | 1 - 2 | 400 | 88 | ||
XM_021476263.1 | XP_021331938.1 | 1 - 2 | 440 | 88 | ||
GenBank mRNA | BC133930.1 | - | 1 - 2 | 153 | 88 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |