Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082839.1 | NP_001076308.1 | 2 - 3 | 293 | 90 | |
XM_005165174.4 | XP_005165231.1 | 2 - 3 | 580 | 90 | ||
XM_021476262.1 | XP_021331937.1 | 2 - 3 | 577 | 90 | ||
XM_021476263.1 | XP_021331938.1 | 2 - 3 | 580 | 90 | ||
GenBank mRNA | BC133930.1 | - | 2 - 3 | 293 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |