Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dph7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082839.1 | NP_001076308.1 | 8 - 9 | 941 | 81 | |
XM_005165174.4 | XP_005165231.1 | 8 - 9 | 1225 | 81 | ||
XM_009301350.3 | XP_009299625.1 | 9 - 10 | 1244 | 81 | ||
XM_021476262.1 | XP_021331937.1 | 8 - 9 | 1225 | 81 | ||
GenBank mRNA | BC133930.1 | - | 8 - 9 | 941 | 81 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |