Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eomesb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083575.2 | NP_001077044.1 | 2 - 3 | 765 | 73 | |
GenBank mRNA | AB269787.1 | - | 2 - 3 | 766 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |