Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2k8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001130623.2 | NP_001124095.1 | 1 - 2 | 312 | 67 | |
GenBank mRNA | AY772712.1 | - | 1 - 2 | 228 | 67 | |
BC163074.1 | - | 1 - 2 | 312 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |