Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ankdd1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001099248.1 | NP_001092718.1 | 1 - 2 | 176 | 64 | |
GenBank mRNA | BC139890.1 | - | 1 - 2 | 176 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |