Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2aa12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109701.2 | NP_001103171.2 | 4 - 5 | 694 | 75 | |
GenBank mRNA | BC152006.1 | - | 4 - 5 | 694 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |