Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131348.2 | NP_571423.1 | 4 - 5 | 689 | 100 | |
XM_005159278.4 | XP_005159335.1 | 5 - 6 | 969 | 100 | ||
XM_021467581.1 | XP_021323256.1 | 5 - 6 | 1015 | 100 | ||
XM_021467582.1 | XP_021323257.1 | 5 - 6 | 996 | 100 | ||
XM_021467583.1 | XP_021323258.1 | 5 - 6 | 988 | 100 | ||
XM_021467584.1 | XP_021323259.1 | 4 - 5 | 794 | 100 | ||
XM_021467585.1 | XP_021323260.1 | 4 - 5 | 713 | 100 | ||
GenBank mRNA | AF177538.1 | - | 3 - 4 | 549 | 100 | |
AJ249590.1 | - | 4 - 5 | 630 | 100 | ||
BC056588.1 | - | 4 - 5 | 678 | 100 | ||
BC066571.1 | - | 4 - 5 | 689 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |