Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fli1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131348.2 | NP_571423.1 | 6 - 7 | 819 | 82 | |
XM_005159278.4 | XP_005159335.1 | 7 - 8 | 1099 | 82 | ||
XM_021467581.1 | XP_021323256.1 | 7 - 8 | 1145 | 82 | ||
XM_021467582.1 | XP_021323257.1 | 7 - 8 | 1126 | 82 | ||
XM_021467583.1 | XP_021323258.1 | 7 - 8 | 1118 | 82 | ||
XM_021467584.1 | XP_021323259.1 | 6 - 7 | 924 | 82 | ||
XM_021467585.1 | XP_021323260.1 | 6 - 7 | 843 | 82 | ||
GenBank mRNA | AF177538.1 | - | 5 - 6 | 679 | 82 | |
AJ249590.1 | - | 6 - 7 | 760 | 82 | ||
BC066571.1 | - | 6 - 7 | 819 | 82 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |