Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: atp1a3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131684.2 | NP_571759.2 | 11 - 12 | 1918 | 99 | |
GenBank mRNA | AF308599.1 | - | 11 - 12 | 1844 | 99 | |
BC064703.1 | - | 11 - 12 | 1918 | 99 | ||
BC065650.1 | - | 11 - 12 | 1971 | 99 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |