Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: vent | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131700.1 | NP_571775.1 | 1 - 2 | 221 | 127 | |
GenBank mRNA | AF255044.1 | - | 1 - 2 | 221 | 127 | |
AF277077.1 | - | 1 - 2 | 215 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |