Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gad1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194419.1 | NP_919400.1 | 10 - 11 | 1138 | 132 | |
XM_009302103.3 | XP_009300378.1 | 10 - 11 | 1130 | 132 | ||
XM_009302104.3 | XP_009300379.1 | 9 - 10 | 1315 | 132 | ||
GenBank mRNA | AB183390.1 | - | 10 - 11 | 1322 | 132 | |
AF017266.1 | - | 4 - 5 | 359 | 132 | ||
AF042374.1 | - | 4 - 5 | 348 | 132 | ||
BC047851.1 | - | 10 - 11 | 1138 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |