Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gad1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_194419.1 | NP_919400.1 | 2 - 3 | 234 | 63 | |
XM_009302103.3 | XP_009300378.1 | 2 - 3 | 226 | 63 | ||
GenBank mRNA | AB183390.1 | - | 2 - 3 | 418 | 63 | |
BC047851.1 | - | 2 - 3 | 234 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |