Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ivns1abpa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199279.2 | NP_954973.2 | 3 - 4 | 554 | 58 | |
XM_021468374.1 | XP_021324049.1 | 3 - 4 | 493 | 58 | ||
GenBank mRNA | BC046068.1 | - | 4 - 5 | 459 | 58 | |
BC066513.1 | - | 3 - 4 | 522 | 58 | ||
DN860974.1 | - | 1 - 2 | 120 | 58 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |