Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pabpc1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199839.1 | NP_956133.1 | 4 - 5 | 603 | 69 | |
GenBank mRNA | AY398370.1 | - | 4 - 5 | 614 | 69 | |
BC044513.1 | - | 4 - 5 | 603 | 69 | ||
BC165470.1 | - | 3 - 4 | 541 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |