Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: trmt2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199929.1 | NP_956223.1 | 4 - 5 | 1040 | 84 | |
GenBank mRNA | BC045367.1 | - | 4 - 5 | 1040 | 84 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |