Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mylipa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_199983.1 | NP_956277.1 | 4 - 5 | 806 | 109 | |
GenBank mRNA | AY423020.1 | - | 4 - 5 | 806 | 109 | |
AY434450.1 | - | 4 - 5 | 768 | 109 | ||
BC045844.1 | - | 4 - 5 | 794 | 109 | ||
BC065679.1 | - | 4 - 5 | 769 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |