Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn11b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200454.1 | NP_956748.1 | 6 - 7 | 841 | 98 | |
XM_005162157.4 | XP_005162214.1 | 6 - 7 | 879 | 98 | ||
XM_009296835.3 | XP_009295110.1 | 6 - 7 | 1033 | 98 | ||
GenBank mRNA | BC055139.1 | - | 6 - 7 | 841 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |