Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: zfand5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200949.1 | NP_957243.1 | 2 - 3 | 295 | 135 | |
XM_021479140.1 | XP_021334815.1 | 2 - 3 | 336 | 135 | ||
GenBank mRNA | BC050491.1 | - | 2 - 3 | 295 | 135 | |
BC165024.1 | - | 1 - 2 | 196 | 135 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |