Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201034.2 | NP_957328.2 | 4 - 5 | 627 | 96 | |
GenBank mRNA | BC055159.1 | - | 4 - 5 | 572 | 96 | |
CT621066.2 | - | 4 - 5 | 486 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |