Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: phb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201297.1 | NP_958454.1 | 3 - 4 | 286 | 70 | |
XM_005163797.4 | XP_005163854.1 | 3 - 4 | 371 | 70 | ||
GenBank mRNA | BC055384.1 | - | 3 - 4 | 286 | 70 | |
BC065895.1 | - | 3 - 4 | 287 | 70 | ||
KM655719.1 | - | 2 - 3 | 242 | 70 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |