Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: apmap | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212608.1 | NP_997773.1 | 6 - 7 | 814 | 78 | |
GenBank mRNA | BC044505.1 | - | 6 - 7 | 814 | 78 | |
BC067549.1 | - | 6 - 7 | 835 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |