Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212654.1 | NP_997819.1 | 2 - 3 | 278 | 108 | |
GenBank mRNA | BC044373.1 | - | 2 - 3 | 278 | 108 | |
BC066698.1 | - | 2 - 3 | 271 | 108 | ||
BC164031.1 | - | 2 - 3 | 182 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |