Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212654.1 | NP_997819.1 | 6 - 7 | 817 | 125 | |
GenBank mRNA | BC044373.1 | - | 6 - 7 | 817 | 125 | |
BC066698.1 | - | 6 - 7 | 810 | 125 | ||
BC164031.1 | - | 6 - 7 | 721 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |