Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hinfp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213424.2 | NP_998589.2 | 8 - 9 | 1541 | 68 | |
GenBank mRNA | AY648714.1 | - | 8 - 9 | 1541 | 68 | |
BC057479.1 | - | 9 - 10 | 1284 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |