Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: prpf38b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_213489.1 | NP_998654.1 | 4 - 5 | 704 | 68 | |
XM_005171371.4 | XP_005171428.1 | 4 - 5 | 734 | 68 | ||
GenBank mRNA | BC061448.1 | - | 4 - 5 | 704 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |