Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: uqcrc2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001589.1 | NP_001001589.1 | 12 - 13 | 1183 | 86 | |
GenBank mRNA | BC054137.1 | - | 12 - 13 | 1161 | 86 | |
BC071551.1 | - | 12 - 13 | 1183 | 86 | ||
BC097011.1 | - | 12 - 13 | 1142 | 86 | ||
BC152592.1 | - | 12 - 13 | 1161 | 86 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |