Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ctnnb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001889.1 | NP_001001889.1 | 8 - 9 | 1352 | 103 | |
GenBank mRNA | AF329680.1 | - | 8 - 9 | 1352 | 103 | |
BC056276.1 | - | 8 - 9 | 1349 | 103 | ||
BC114257.1 | - | 1 - 2 | 101 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |