Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pbk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002387.1 | NP_001002387.1 | 2 - 3 | 126 | 72 | |
GenBank mRNA | BC075869.1 | - | 2 - 3 | 126 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |