Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cryba1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002410.1 | NP_001002410.1 | 4 - 5 | 366 | 94 | |
XM_005157649.4 | XP_005157706.1 | 4 - 5 | 398 | 94 | ||
GenBank mRNA | BC076170.1 | - | 4 - 5 | 366 | 94 | |
DQ000462.1 | - | 4 - 5 | 366 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |