Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: otub1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002411.1 | NP_001002411.1 | 3 - 4 | 243 | 76 | |
GenBank mRNA | BC076168.1 | - | 3 - 4 | 243 | 76 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |