Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lyrm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002506.1 | NP_001002506.1 | 2 - 3 | 227 | 95 | |
GenBank mRNA | BC076293.1 | - | 2 - 3 | 227 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |