Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cdc34b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002688.1 | NP_001002688.1 | 1 - 2 | 646 | 129 | |
GenBank mRNA | BC076537.1 | - | 1 - 2 | 646 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |